Symmetrical Round Jaws Buret Clamps
Supplier: GSC International
4-BC37-UC
470005-372EA 14.4 USD
470005-372
Symmetrical Round Jaws Buret Clamps
Clamps General Purpose Clamps
Hold round objects from 9.5–35mm in diameter with this versatile clamp.
The jaws can be rotated 360° and may be locked in place at any angle. The clamp can be quickly attached to support rods up to 16mm in diameter. Overall length is 16cm.