Ripple Tank/Wave Generator
Supplier: Electro-Technic Products
40003
470005-286EA 99.29 USD
470005-286
Ripple Tank/Wave Generator
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Designed Specifically for Use with an Overhead Projector
  • Easy to set up
Your whole class will be able to clearly see your demonstrations of propagation, reflection, diffraction, frequency-wavelength-velocity relationships, and interference and phase differences when you place the ripple tank on your overhead projector. It comes with a clear plastic ripple tank, wave generator, and a variety of barriers. The apparatus attaches easily to a ring stand, available separately. The wave generator requires a variable low voltage DC power supply, or a 1.5 V dry cell with rheostat, also available separately.