Optical Voice Link Kit
Supplier: Industrial Fiber Optics
IF OVL10K
470004-994EA 106 USD
470004-994
Optical Voice Link Kit
Educational Materials Physics Educational Materials Optics Learning Activities
The Optical Voice Link Kit is the ideal introduction for those students first learning about the marvels, mysteries and science of light transmission through optical fiber.
Students will be fascinated by hearing their own voices, after it has been converted into light and then coupled into, through and out of an optical fiber. This kit is suitable for science projects, home projects, short audio fiber optic curriculums for schools, and inexpensive classroom demonstrations.
No prior fiber optics experience, special tools or training are needed for students to be able to build, use and enjoy the multiple applications of this kit.
Some basic soldering is required. Three meters of optical fiber is included but the kit is expandable to 20 meters.
Delivery information: Includes: printed wiring boards, switches, electronics, microphone, 8-ohm speaker, 3 meters of plastic optical fiber, 32-page instructional manual.