Sunspotter Keplerian Telescope
Supplier: Science First
654-0145
470004-984EA 529 USD
470004-984
Sunspotter Keplerian Telescope
Telescopes
The Sunspotter Keplerian Telescope lets users safely view the light of the sun.
  • Length: 16"
  • Width: 15"
  • Height: 6"
  • Altitude-Azimuth Design
  • Also for Use with Eclipses
This wooden, folded-Keplerian telescope is safe and easy to use. By using a series of mirrors, the telescope projects a 85 mm (3½”) image of the sun onto a white viewing screen several students can see at once. They can trace the changing positions of sunspots and compare them on a daily basis. The kid-friendly telescope has an altitude-azimuth design and can be used to observe eclipses as well.
Features include a 700 mm (27½”) FL objective lens, 61.7 mm (2½”) objective aperture, 12.5 mm (1/2”) fully-coated FL Plössl field lens, and 0.75° field of view.