Helium-Neon Laser
Supplier: Industrial Fiber Optics
IF-HN08M
470004-978EA 1008.1 USD
470004-978 470102-646
Helium-Neon Laser
Laser Pointers
The Helium-Neon Laser is a strong laser source for holography and interferometry.
  • Wavelength: 632.8 nm
  • AM/FM Radio Transmission
  • Solid-State Electronic Design
  • Tamper-Resistant Screws for Safety
  • Ideal for Holography and Interferometry
Helium neon (He Ne) lasers have wavelength stability (632.8 nm) and long coherence length. Features include AM/FM radio transmission, diffraction, and single and double slit experiments. The versatile lasers are stored in attractive blue acrylic casing.
The lasers have solid-state electronic design, a fool-proof mechanical beam stop, tamper-resistant screws for safety, threaded mounts for holding lenses; filters; and diffraction gratings, and rubber feet for adhesion on slick lab surfaces.