Heavy Duty Rock Tumbler, 15 lbs. Capacity
Supplier: TRU-SQUARE METAL PRODUCTS
140
470004-964EA 499.95 USD
470004-964
Heavy Duty Rock Tumbler, 15 lbs. Capacity
Educational Materials Earth Science Educational Materials Minerals and Rocks Learning Activities
Turn rough rocks into beautifully polished gemstones. This professional quality tumbler features a 15 pound capacity steel hexagonal barrel with removable rubber liner.
  • Size: 13½×12¾×11½"
  • Weight: 15.5 lbs.
It can be used for a variety of tumbling and mixing operations, and will polish brass and other metals as well as rocks. The 115 V heavy duty motor is fan-cooled and overload protected.