Longitudinal Wave Demonstration
Supplier: Avantor
470004-950EA 116.09 USD
470004-950
Longitudinal Wave Demonstration
Educational Classroom Kits and Activities
This inexpensive model, developed by Tim Ernest, lets you create longitudinal/compression waves by simply pushing a Slinky® along a track.
  • Grades 6 to 9
  • Length: 152.4 cm
It can be used in studies of waves, and sound to demonstrate compression, rarefaction, amplitude, wavelength, echoes, volume and pitch.
Delivery information: Includes instructions.