Solar Electricity Kit
Supplier: Solar Made
SK-50
470004-940EA 27.05 USD
470004-940
Solar Electricity Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
With this Solar Cell Kit, you can harness the power of the sun.
  • Experiment with natural and artificial light
  • Use solar power to run several devices
  • Includes an instruction manual
This Solar Energy Kit consists of an encapsulated solar cell array, a miniature motor, a wind propeller, an optical illusion disc and an instruction manual.
Students can create a solar desk fan and a "virtual spiral" device, study the impact of artificial light versus sunlight and conduct numerous other experiments using the solar cell.