Biology And Chemistry Of Soil Kit
Supplier: LAB-AIDS INC (KITS) SE
32
470004-612EA 176.05 USD
470004-612
Biology And Chemistry Of Soil Kit
Standards Environmental Analysis Standards
Soil Feeds Us And Sustains Us.
  • Conduct A Thorough Evaluation Of Soil
  • Identify The Type And Variety Of Micro-Organisms (Soil Microbes)
  • Time Required: One 50 Minute Class Period
Time to play in the dirt! Students use ordinary soil samples to conduct a thorough evaluation of soil. Simple, easy-to-follow techniques and staining procedures are introduced early in this series of activities. After performing a series of activities, students are asked to draw conclusions concerning a soil’s ability to support plant growth. All tests are performed using either the patented Chemplate® or calibrated sample tubes.
Included:
Teacher’s Guide with SDS
Student Worksheets and Guides
Chemplates®, LAB-AIDS®
Cups, plastic
Plastic slides
Tubes, calibrated
Barium Chloride Solution, Drop control bottle
Phenolic Rose Bengal, Drop control bottle
pH Soil Indicator Solution, Drop control bottle
Potassium Dichromate Solution, Drop control bottle