Ward's® Regulation of Gene Expression Lab Activity
Supplier: Avantor
470004-542KT 74.99 USD
470004-542
Ward's® Regulation of Gene Expression Lab Activity
Educational Classroom Kits and Activities
Simple Experiment with Easy-to-See Results
  • Dramatically demonstrate inducible phenotype
  • Materials for 6 lab groups
  • Time Required: ~60 minutes
With a simple experiment, students investigate how gene expression can be controlled by regulatory enzymes and environmental factors. When the bacterial gene is expressed and an enzyme is produced, the samples turn a brilliant shade of red. The kit comes with enough materials for six setups, a teacher’s guide, and student copymaster.
Canada: Containment Level 2 – PHAC License required.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.