3B Scientific® Stomach Model
Supplier: American 3B Scientific
1000303
470004-536EA 487.1 USD
470004-536
3B Scientific® Stomach Model
Models Human Biology Models
Shows Relationship With Duodenum And Pancreas
  • Shows Ulcer Erosions At Different Magnifications
  • 3 Year Manufacturers Warranty
  • Includes 3B Smart Anatomy
This life-size stomach model has a portion of the duodenum and pancreas attached to show how the organs work together in the human system. Your class can study the interrelationships with the model as a whole, then separate the three parts to study each one individually. The stomach and pancreas are dissected to show their inner structures; the pancreatic ducts are shown in relation to the duodenum. Vessels and nerves are also shown, as are the layers of the stomach wall. The three-piece model is mounted on a base and can be removed for closer study.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.