3B Scientific® Giant Eye In Bony Orbit
Supplier: American 3B Scientific
1000257
470004-514EA 609.25 USD
470004-514
3B Scientific® Giant Eye In Bony Orbit
Models Human Biology Models
This model covers all the important structures of the human eye.
  • Three Times Life Size
  • Eye Sits Inside Bony Orbit Base
  • Three-Year Manufacturer's Warranty
  • Removable Parts
  • Includes 3B Smart Anatomy
Removable parts of the model include the upper half of the sclera with cornea and eye muscle attachments, both halves of choroid with iris and retina, eye lens, vitreous humor, eyelid, lachrymal system, and anatomical features around the eyeball.
The eye sits inside a bony orbit base.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.