Ward's® Student Hand Microtome
Supplier: Avantor
470004-504EA 49 USD
470004-504 470147-178
Ward's® Student Hand Microtome
Microtomes and Microtome Assemblies Microtomes
Explore section slide making
  • Create your own slides with your own specimens
  • Safety features prevent injury
Our unique, easy-to-use microtome features a built-in safety flange to protect fingers from cuts. It includes wax embedding granules, razor knife, five blades, and instructions.
2" dia. platform, 2" length