Chromatography Paper Strips
Supplier: Precision Laboratories
CHROM-50-6X75
470004-492PK 6.25 USD
470004-492
Chromatography Paper Strips
Filters Filter Paper Chromatography Paper
Perfect for your paper chromatography needs.
  • Package of 50
This chromatography paper is supplied in strips rather than rolls that are apt to stretch in order to assure the most even distribution of material being separated for more accurate results.