Basic Hand-Held Stream Flowmeter
Supplier: GEOPACKS
ZMFP51
470004-378EA 422.2 USD
470004-378
Basic Hand-Held Stream Flowmeter
Flow Meters
Lets You Control Location and Depth of Measurements
Using the three included elevation rods, you can measure velocity at fixed heights from 4–19" above the stream bed. The digital LCD meter is stored in a protective carrying pouch, and comes with a sturdy impeller stick that disassembles into four sections for easy storage, an instruction manual with laminated velocity calibration sheets and data sheets, and a compact weather-resistant case that conveniently holds the flowmeter, all additional parts, and two AAA batteries. Size of carrying case: 8" x 14" x 7". Total pole length: 3 1/2', extendable to 5' with an overall 7' lead. Weight: 2 lbs.