Comprehensive Water Monitoring Kit
Supplier: LaMotte
5848
470004-240EA 357.28 USD
470004-240
Comprehensive Water Monitoring Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Most factors can be tested 100 times each with the Comprehensive Water Monitoring Kit.
  • No hazardous compounds needed for testing
  • Monitoring kit manual is in english and spanish
  • Single-factor test kits
  • Enough TesTabs to test nine factors 100 times each
  • Aligned to NGSS 4-ESS2-1, 4-ESS2-2, MS-LS2-1, MS-LS2-5, MS-ESS3-3, HS-LS2-7, HS-ESS2-5, HS-ESS3-4
Using TesTabs, which are safe, nonhazardous reagent tablets, students can easily test for a variety of factors without risk. Developed by the Global Rivers Environmental Education Network, each monitoring kit comes with a bilingual (English and Spanish) manual.
The kit include enough TesTabs to test nine factors. Test factors include benthic macroinvertebrates, biochemical oxygen demand, coliform bacteria, nitrate, dissolved oxygen, pH, phosphate, turbidity, and temperature change. The kit have enough material for 100 tests each (44 coliform tests; unlimited benthic invertebrate, turbidity, and temperature change tests).