The Greenhouse Effect: Exploring the Theory Kit
Supplier: Avantor
470004-222KT 103.09 USD
470004-222
The Greenhouse Effect: Exploring the Theory Kit
Models Meteorology Models
Here's an easy way to explore the greenhouse effect theory.
  • Directly measure temperature differences
  • Includes teacher's guide
  • Grades 5 - 8
Students create two identical soil-filled environments, except one is open and one is closed. Both environments are exposed to the same conditions.
By taking periodic temperature measurements and graphing their data, students can clearly see differences in rates of heating and cooling in the two bottles. Developed by Freddie Faber. Grades 5 - 8.
Includes materials for eight student groups: thermometers, soil, rubber stoppers, soda bottles, X-acto knife, a teacher's guide and student instructions.
Please note: Some assembly is required. A balance or scale and a reflector lamp with at least a 100 W bulb are recommended for each lab station but are not included.