Denoyer-Geppert® Comprehensive Spinal Cord Model
Supplier: Denoyer-Geppert Science
0165-00
470004-200EA 490.2 USD
470004-200
Denoyer-Geppert® Comprehensive Spinal Cord Model
Models Human Biology Models
Diagramed and color-coded for complete study.
  • Mounted on Base
  • Enlarged 6 Times
The upper thoracic spinal cord is rendered in longitudinal and cross section and enlarged six times to show dorsal and ventral nerve roots with ganglia as well as the blood supply. Each type of nerve has been painted a different color: afferent visceral nerves, green; afferent somatic nerves, yellow; efferent visceral nerves, red; and efferent somatic nerves, black.
The model is mounted on a base, which features a circuitry diagram.