BeeSpi Timer
Supplier: NADA SCIENTIFIC LTD.
N99-S77-1321
470004-180CT 201.45 USD
470004-180
BeeSpi Timer
Educational Materials Physics Educational Materials Mechanics Learning Activities
This versatile timer with infrared sensors can be used to measure the speed of a moving object.
  • Hand-sized
  • Functions Include Lap Time and Cumulative Lap Time
  • Infrared Sensors
  • Easy-To-Read LCD Screen
  • For Measuring the Speed of a Moving Object
It can also determine the earth’s gravity of falling objects, measure the acceleration of free-falling objects, determine the terminal velocity with the addition of masses, and more. Use with an inclined plane to measure the relationship between the angle of incline and the acceleration of a moving object.
Functions include speed measuring, lap time, and cumulative lap time. The large LCD display is easy to read. It requires two AAA batteries.
Ordering information: Two AAA batteries are required but not included.