Decade Resistance Box
Supplier: Shiv Dial Sud & Sons
P50575
470004-130CS 1439.28 USD
470004-130
Decade Resistance Box
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This box changes resistances in a laboratory circuit.
  • Resistance Range: 0 to 1,111,110 Ohm
  • Maximum Power Dissipation: 1.0W
  • Universal Binding Posts Mounted on Front Panel
  • Circuit Connects to Any Kind of Attachment with Ease
  • For Changing Resistances in Laboratory Circuits
Universal binding posts are mounted on front panel for easy connections to the circuit with any kind of attachment. 
The resistance range is 0 to 1,111,110 Ohm, and the maximum power dissipation is 1.0W.