Leaf Identification Kit
Supplier: Young Naturalist
201
470004-106KT 65.65 USD
470004-106
Leaf Identification Kit
Educational Materials Biology Educational Materials Plant Learning Activities
Real leaves for comparison.
Thirty-nine different tree leaves are supplied, including maple, oak, elm, ash, and others; students compare their sizes and shapes as well as their veins, margins, and position on twigs as an important step toward learning how to identify trees. This kit also teaches seeds’ different means of transport. It comes with a teacher’s guide and 61-page Tree Finder book.