Student gratings
Supplier: United Scientific Supplies
DFG600 DFG300 DFG100
470004-102CS 816 USD
470004-102 470158-406 470158-408
Student gratings
Educational Materials Physics Educational Materials Optics Learning Activities
Quality gratings perfect for your Optics Needs
  • Viewing area measures approx. 45x30mm
Quality transmission gratings protected by two glass plates, 60x45mm.