Rapid Radish Seeds
Supplier: Avantor
Rapid Radish seeds are selectively bred to germinate and grow quickly, giving you fast, accurate results.
The large seeds are easy to handle and also emerge easily from the soil, with a high germination rate. In addition, the plants, which remain hardy throughout their life cycle, feature large flowers that allow for the study of various structures.This item is NOT irradiated.