Munsell® Soil Color Chart
Supplier: Peco Sales
77321
470003-876EA 406.55 USD
470003-876
Munsell® Soil Color Chart
Charts
Features Over 300 Color Chips
Approximately 322 chips are permanently mounted on charts in a loose-leaf binder to make it easy to compare soil color, rocks, and archaeological specimens. The chart also includes two Gley Charts for easier comparison. Size: 7 5/8" x 5 5/8".