Hanna Instruments Marine Test Kit
Supplier: Avantor
470003-780KT 336 USD
470003-780
Hanna Instruments Marine Test Kit
Environmental Test Kits
This chemical test kit for the determination of six parameters commonly monitored for aquaculture testing: alkalinity, carbon dioxide, hardness, dissolved oxygen, pH, and salinity. This kit is equipped with all the necessary reagents and equipment to perform over 100 tests for each parameter. pH is obtained with Hanna's pH tester, which guarantees greater accuracy and a longer life than traditional litmus paper.
  • Pre-made reagents for ease of use
  • All reagents marked with expiration date and lot number for traceability
  • Portable carrying case for easy transport
It is a chemical test kit that uses titration and direct measurement to measure six parameters common in aquaculture testing: alkalinity, carbon dioxide, hardness, dissolved oxygen, pH, and salinity. It is supplied with all of the necessary reagents and equipment to perform each analysis, and all reagents are individually available as they run out.
Ordering information: Replacement reagents need to be purchased seperatly.