The Remagnetizer
Supplier: Master Magnetics
DM-RM
470003-722EA 117.9 USD
470003-722
The Remagnetizer
Magnets
Lightweight and reliable
  • Instructions are included
  • CSA approved
  • Size: 10 x 13 x 7cm
Save yourself time and money by magnetizing your Alnico horseshoe and bar magnets when they become weak. This handy tool contains two permanent neodymium magnets to simply and quickly recharge your magnets. The Remagnetizer can be mounted to a wall or table and requires no electricity. It has a durable aluminium frame and steel base, and weighs only 1.38 kg (3.05 lbs.).