Static Wand
Supplier: Avantor
470003-682EA 45.79 USD
470003-682
Static Wand
Educational Classroom Kits and Activities
Illustrate Electrostatic Propulsion and Repulsion
  • The set includes a static wand, four static bands, rabbit fur, and teacher’s guide
Rub the rabbit fur over the wand to create a charge, then hold the wand beneath the static band and watch it hover in mid-air, even as you walk around the room.