Celestron NexStar 4 SE Maksutov-Cassegrain Telescope
Supplier: Celestron International
11049
470003-600EA 911.19 USD
470003-600
Celestron NexStar 4 SE Maksutov-Cassegrain Telescope
Telescopes
Celestron NexStar 4 SE Maksutov-Cassegrain Telescope is ready to use in minutes.
  • Custom Database
  • Internal Flip Mirror
  • StarPointer Finderscope
  • Starbright XLT High Transmission Coatings
  • Computerized Altazimuth Mount
Combining the heritage of the original orange-tube telescope with state-of-the-art features, this computerized telescope features a custom database that lists the most popular deep-sky objects. It also includes an internal flip mirror for convenient viewing and a StarPointer Finderscope to assist with alignment and locating objects. StarBright XLT high transmission coatings are standard on this model.
The camera-control feature and shutter-release cable allow you to capture a series of exposures remotely when using a digital SLR camera.
Ordering information: This telescope includes SkyAlign, flash upgradeable hand control software, a 40,000 object database, an internal battery compartment to prevent cord wrap during use, a computerized altazimuth mount, and a steel tripod. It requires eight AA batteries, not included.