Student Thermometer
Supplier: SI Manufacturing
95570
470003-560EA 11.16 USD
470003-560
Student Thermometer
Thermometers Analog Thermometers
Student thermometer ideal for all grade levels. Alcohol thermometer recessed in molded plastic to prevent breakage. Unlike non-plastic thermometers, it can be immersed in hot liquids without danger of burning students hands.