Pocket Laser Pointer

Supplier: THE BEST CONNECTION INC
4012
470003-492EA 47.95 USD
470003-492
Pocket Laser Pointer
Laser Pointers
Class IIIa red laser pointer for optics experiments.

  • Classically designed laser pointer
  • Range of over 500 yards
  • Maximum power: 5 mW (Class IIIa)
  • Approximate wavelength: 650 nm
  • Made of aircraft aluminum
  • Dimensions: 5³/₈×⁷/₁₆"
  • Weight: 4.7 oz.

Ideal for indoor and outdoor use; for presentations and instruction.

Ordering information: Includes 2 AAA batteries and a plastic gift/storage box.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR