Vernier® Microphone
Supplier: Vernier Software & Technology
MCA-BTA
470002-962EA 54.99 USD
470002-962
Vernier® Microphone
Sound Meters
Vernier Microphone helps students learn about waveforms of sound.
  • For Displaying and Studying Waveforms
  • Typical Maximum Frequency: 10,000 Hz
  • Power: 1.45 mA At 5 VDV
  • Ideal for Speed of Sound Experiments
  • For Educational Use Only
The Microphone can be used to display and study the waveforms of sounds from voices and musical instruments.
It is also great for speed of sound experiments.