Vernier® 3-Axis Accelerometer
Supplier: Vernier Software & Technology
3D-BTA
470002-946EA 104.99 USD
470002-946
Vernier® 3-Axis Accelerometer
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Analyze separate components of complex accelerations.
  • Measure up to 5 G
  • Compact design
The 3-Axis Accelerometer consists of three –5 to +5 g accelerometers mounted in one small block. Using the appropriate data collection hardware and software, you can graph any of these components, or calculate the magnitude of the net acceleration. The 3-Axis Accelerometer can be used for a wide variety of experiments and demonstrations, both inside the lab and outside.
Power: 30 mA at 5 VDC
Frequency Response: 0–100 Hz
For educational use only