Easy-Read™ Thermometer
Supplier: Bel-Art Products
B60300-0100
470002-856EA 26.35 USD
470002-856
Easy-Read™ Thermometer
Thermometers Analog Thermometers
Great for All Grade Levels.
  • Enviro-Safe black liquid against lead-free, yellow back glass for easy readability and fewer reading errors
  • Manufactured and tested by our USA quality team
  • Traceable to NIST
  • Partial immersion 5 cm (2")
  • Length 20 cm (7.9")
Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability, non-roll fitting and individual serial number.