3B Scientific® Introductory Disarticulated Skeleton
Supplier: American 3B Scientific
1020157
470002-720EA 727.5 USD
470002-720
3B Scientific® Introductory Disarticulated Skeleton
Models Human Biology Models
This skeleton is perfect for introducing individual bones of human form to students.
  • Full-Size Replica
  • Calvarium Sectioned for Close-Up View
  • Bones Are Separate Except for Hands and Feet
  • Hinged Jaw
  • Includes 3B Smart Anatomy
The skeleton’s bones are full-size replicas, and all bones are separate with the exception of both hands and feet. One hand and foot are fully articulated on sturdy wire while the other hand and foot are articulated on nylon to demonstrate natural movement.
The skull’s calvarium is sectioned for a close-up view, and the jaw is hinged. The sternum’s intercostal cartilage is attached.
Accessories information: Ward's Human Osteological Preparations brochure is available for download. For information about 3B Smart Anatomy, please see the document attached on this page.