Amoeba Model
Supplier: The Science Source
B 136
470002-680EA 477.4 USD
470002-680
Amoeba Model
Models Zoology Models
View The Same Structures As Living Organisms Without A Microscope
The 1,000X translucent model realistically portrays an amoeba in motion, with prominent pseudopods, representing natural movement. Sectioned to show internal cell details, the center of the amoeba clearly displays the nucleus, food vacuoles containing diatoms and bacteria, a fat droplet, contractile vacuole, and granular and clear zones of cytoplasm. The one-piece model is mounted on a base and includes a key, identifying 11 structures.