Soil Formation Lab Kit
Supplier: The Science Source
2600
470002-488EA 200.95 USD
470002-488 470200-090
Soil Formation Lab Kit
Educational Materials Environmental Educational Materials Environmental Learning Activities
Explore the science behind soil formation with this hands-on lab kit, ideal for environmental science and geology education.
  • Interactive lab kit – Demonstrates the stages of soil formation through simple, engaging experiments
  • Non-Hazardous materials – Safe for classroom and group use with easy-to-follow instructions
  • Teacher's guide included
  • STEM learning tool – Ideal for teaching earth science, geology, and environmental science
The Soil Formation Lab Kit offers an interactive experience for students to explore soil creation and distinguish between soil and sediment. Through experiments with various materials and environmental factors, students can observe how weathering, erosion, and biological activity contribute to soil development. Ideal for earth science classrooms, this kit provides hands-on learning about soil composition, its role in ecosystems, and the factors influencing soil quality and formation.
Additionally, students will understand the critical importance of soil, despite its thin layer in most parts of the world. They will start with the basics, learning how soil is formed by making their own 'rock' from carbonates, simulating acid rain effects with vinegar and water, observing the role of plant life in soil formation, and distinguishing between finely crushed rock particles and true soil. The kit includes materials for 15 setups, a teacher's guide, and instructions.