Ward's® Exploring Physical Properties of Minerals Lab Activity
Supplier: Avantor
470002-460EA 296.79 USD
470002-460 470328-402
Ward's® Exploring Physical Properties of Minerals Lab Activity
Educational Classroom Kits and Activities
Students Master Investigative Skills by Testing Mineral Properties.
  • Teach students about properties of minerals
  • Teacher and student guides included
  • Aligned to NGSS standards (MS/HS: ESS2, ESS3)
Eight common minerals are supplied to test hardness, color, streak, luster, specific gravity, and other characteristics. Each sample has been carefully selected to ensure that students can readily recognize key physical properties and achieve consistent test results. Each kit contains a streak plate, copper strip, glass plate, nail, magnifier, and graduated cylinder for physical tests, as well as a teacher’s guide and worksheet copymasters.
Minerals: calcite, feldspar, galena, graphite, hematite, muscovite, pyrite, and quartz.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.