Drosophilia Cryolizer-Anesthetizer
Supplier: Kemtec
2-800
470002-086EA 71.8 USD
470002-086
Drosophilia Cryolizer-Anesthetizer
Animal Supplies Animal Anesthesia Systems
This plastic easy-to-use cooling unit immobilizes flies for over thirty minutes.
  • Aanesthetizes without sterilizing or killing specimens
  • With Petri dish: Placed in a freezer, it cools flies to immobilize them
Dia. 3½"; height 3½"