SP Bel-Art Safety Waste Jug, Bel-Art Products, a part of SP
Supplier: Bel-Art
H11919-0000_SE
470002-024EA 284.55 USD
470002-024
SP Bel-Art Safety Waste Jug, Bel-Art Products, a part of SP
Safety Cans
For safe, in-plant handling and temporary storage of spent solvents or other liquid wastes awaiting proper disposal.
  • Flared top on snap-on cap helps retain fumes
  • Translucent
  • HDPE jug construction
For easy filling, the screw-on top has a funnel-like flare, 12 cm (43/4") in diameter. A snap-on cap seals the flared top to retain fumes. Translucent, high-density graduated polyethylene jug.