Four-Channel Timer
Supplier: General Tools Manufacturing
TI388
470001-920EA 52.25 USD
470001-920
Four-Channel Timer
Timers Digital Timers
This timer features a clock and four independent channels that count up or down from any interval from one second to 100 hours — each with its own distinctive alarm.
It will automatically count up after the countdown reaches zero. It features a stand, pocket clip, magnet, and a quartz crystal accurate clock display. It runs on a 1.5 V battery, included. Size: 2⁷/₈L×2¹/₂W×⁹/₁₆"H.