Vernier® Respiration Monitor Belt
Supplier: Vernier Software & Technology
RMB
470001-866EA 62.99 USD
470001-866
Vernier® Respiration Monitor Belt
Pressure Sensors
The respiration monitor belt is used with our gas pressure sensor to measure human respiration.
  • Respiration rate calculation is only supported in Logger Pro and LabQuest App.
Simply strap the belt around your chest, then pump air into the belt with the hand bulb. You can then monitor the pressure associated with the expansion and contraction of your chest during breathing. (Requires the Gas Pressure Sensor.)