Vernier EKG Sensor
Supplier: Vernier Software & Technology
EKG-BTA
470001-862EA 178.99 USD
470001-862
Vernier EKG Sensor
Electrodes Other Electrodes
Measure electrical signals from muscle contractions, or make EKG tracings and surface EMG recordings.
  • Wired connection with BTA plug
  • ~1.00 V (±0.3 V) offset
  • 1 mV body potential/1 V sensor output gain
The EKG Sensor measures cardiac electrical potential waveforms (voltages produced during the contraction of the heart). It can be used to make standard 3-lead EKG tracings to record electrical activity in the heart, or to collect surface EMG recordings to study contractions in muscles in your arm, leg or jaw.
Ordering information: LabQuest® (2, 3, Mini) interface is required but not included.