Magnetic Compass with Beveled Top
Supplier: United Scientific Supplies
CWR045-W
470001-824EA 6.45 USD
470001-824
Magnetic Compass with Beveled Top
Compasses
This simple, student-grade model is not dampened and measures in 2° increments.
  • It is brass-colored and has a ring for attaching a lanyard
  • Great price makes it ideal for outfitting large classes
Size: 2 Ø.×1/2"