Sponge Spicules
Supplier: Avantor
470001-768PK 18.95 USD
470001-768
Sponge Spicules
Specimens Preserved Specimens Preserved Animal Materials
Formalin preserved sponge spicules. Available in vials.
  • Invertebrate anatomy
  • High quality preservation
Spicules, minute structures of calcium carbonate or silicon dioxide, comprise the supporting “skeletons” of many sponges. When the soft tissue of the sponge dissolves, it leaves an assortment of spicules of many different forms.