Preserved Congo Eel
Supplier: Avantor
470001-586EA 37.99 USD
470001-586
Preserved Congo Eel
Specimens Preserved Specimens Preserved Animal Materials
Formalin preserved congo eel. Available in vacuum packs.
  • Amphibian anatomy
  • High quality preservation
The congo eel (Amphiuma sp.) is not an eel at all but a large aquatic salamander. It is the only surviving genus within the family Amphiumidae and is often mistaken for an eel or snake because of its long, slim body type and its small vestigial legs that are not readily obvious. Because of its morphological differences from other salamanders, it makes a great specimen for amphibian comparative anatomy studies. Size: 24" or longer.
Great alternative to Necturus