Ward's® Fish Scale Set
Supplier: Avantor
470000-984KT 54.89 USD
470000-984
Ward's® Fish Scale Set
Specimens Preserved Specimens Preserved Animal Materials
Formalin Preserved
Examine the different types of external structure unique to fish. Contents: ten each of placoid, ganoid, ctenoid, and cycloid scales. The set of four is supplied in separate jars.