Ward's® Fungi Kingdom Survey Set
Supplier: Avantor
470000-952EA 36.95 USD
470000-952
Ward's® Fungi Kingdom Survey Set
Specimens Preserved Specimens Preserved Fungi Materials
Formalin Preserved
Study the morphological differences in the fungi kingdom and a lichen which shows the symbiotic relationship between fungi and algae. Supplied in separate, labeled jars or vials. Contents: Zygomycete (Rhizopus) in vial, Basidiomycete (Mushroom) in jar, Ascomycete (Peziza) in vial, Deuteromycete (Penicillium) in vial, Mycophycophyte (Lichen) in vial. Set of five.