Preserved Purple Sea Urchin
Supplier: Avantor
470000-770PK 35.99 USD
470000-770
Preserved Purple Sea Urchin
Specimens Preserved Specimens Preserved Animal Materials
Formalin preserved purple sea urchins. Available in pails.
  • Invertebrate anatomy
  • High quality preservation
Arbacia punctulata or Strongylocentrotus purpuratus; genus and species dependent on availability.
Characterized by complex feeding structures with five calcareous protractable teeth (Aristotle’s lantern).