Introduction to Plant Tissue Culture Kit
Supplier: Edvotek
908-A
470000-012EA 175.9 USD
470000-012
Introduction to Plant Tissue Culture Kit
Cell Culture Media
Cutting-Edge Experimentation In Your Classroom
  • Convenient Ready-To-Load samples
  • Connection to stem cell biology
  • Open or guided inquiry-based experiment
  • Time required: ~60 minutes with subsequent growth over several weeks
Though a highly controversial area of biotechnology, genetic modification of plants is an ever growing area of research. These experiments all begin by establishing plant cells in culture, which involves de-differentiating plant cells to form plant "stem cells". In this experiment, students will establish cell cultures of African violets from leaves. Plant growth regulators will then be used to encourage root growth, eventually producing a mature plant. African violets are available separately.
No. of Lab Groups: 10