Bioact® Cleaner, MicroCare
Supplier: MicroCare
MCC-EC7MP
46620-666EA 2623.15 USD
46620-666
Bioact® Cleaner, MicroCare
Soldering Supplies Flux Removers
Based on biodegradeable citrus terpenes derived from Florida oranges, this solvent aggressively cleans water-soluble fluxes and pastes.
  • Nontoxic
  • Combustible
  • Ozone-safe
Very effective on rosin, synthetic, and no-clean fluxes. Bioact® replaces traditional CFC solvents, does not need a water rinse, and has a pleasant citrus scent.